missing translation for 'onlineSavingsMsg'
Learn More

Mercurochrome, 24-27% Mercury

Product Code. 10553171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553171 25 g Glass bottle 25g
10308670 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 10553171 Supplier Thermo Scientific Acros Supplier No. 190110250
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 129-16-8 | C20H8Br2HgNa2O6 | 750.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-16-8
Molecular Formula C20H8Br2HgNa2O6
Molecular Weight (g/mol) 750.65
MDL Number MFCD00013081
InChI Key QJJYLIJNUFJILQ-UHFFFAOYSA-N
Synonym Merbromin, 2, 7-Dibromo-4-hydroxymercurifluoresceine disodium salt
PubChem CID 54598522
IUPAC Name sodium;[2,7-dibromo-9-(2-carboxyphenyl)-3-hydroxy-6-oxoxanthen-4-yl]mercury;hydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C=C3OC4=C(C(=C(C=C24)Br)O)[Hg])Br)C(=O)O.O.[Na+]

Specifications

Melting Point >300°C
Color Green
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 5934
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol,acetone,ether
Formula Weight 750.65
Percent Purity 24 to 27% (Hg), 18 to 22.5% (Br2)
Physical Form Glimmering Scales, Granules or Crystals
Chemical Name or Material Mercurochrome
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercurochrome, 24-27% Mercury
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.