missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) Nitrate Dihydrate, ≥97%, Honeywell™

Product Code. 15696700 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15696700 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15696700 Supplier Honeywell Research Chemicals Supplier No. 680265250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 70°C
CAS 7782-86-7
Color White
Physical Form Crystalline
Quantity 250 g
Molecular Formula H4Hg2N2O8
Linear Formula Hg2(NO3)2 · 2H2O
MDL Number MFCD00150813
UN Number UN1627
InChI Key LSABZDVKJBWCBE-UHFFFAOYSA-N
SMILES O.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name dimercury(1+) dihydrate dinitrate
Molecular Weight (g/mol) 561.22
Formula Weight 561.22g/mol
Percent Purity ≥97%
Packaging Poly bottle
Density 4.78g/mL at 25°C
Chemical Name or Material Mercury(I) nitrate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.