missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) sulfate, 97%, Thermo Scientific Chemicals

Product Code. 10589423 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
100 g
Packaging:
Plastic bottle
missing translation for 'unitSize'
100g
5g
This item is not returnable. View return policy

Product Code. 10589423

missing translation for 'mfr': Thermo Scientific Acros 389920050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-36-0
Molecular Weight (g/mol) 497.24
MDL Number MFCD00016138
InChI Key MINVSWONZWKMDC-UHFFFAOYSA-L
Synonym mercurous sulfate, dimercury sulfate, dimercury sulphate, mercury i sulfate, sulfuric acid, dimercury 1+ salt, unii-pi950n9dys, mercurous sulphate, sulfuric acid, dimercury 1++ salt, pi950n9dys
PubChem CID 24545
IUPAC Name mercury(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Hg+].[Hg+]

Specifications

Color Beige or White-Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 96.0 to 104% (Iodimetry)
Molecular Formula Hg2O4S
Linear Formula Hg2SO4
Merck Index 15, 5965
Solubility Information Solubility in water: 0.06g/100mL (25°C). Other solubilities: soluble in diluted nitric acid
Formula Weight 497.24
Percent Purity 97%
Packaging Plastic bottle
Chemical Name or Material Mercury(I) sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(I) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.