missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) acetate, 98+%

Product Code. 11434094
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434094 500 g 500g
11414094 25 g 25g
11424094 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434094 Supplier Thermo Scientific Alfa Aesar Supplier No. A12478.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1600-27-7 | C4H6HgO4 | 318.68 g/mol

Mercury(II) acetate is used as a reagent to prepare organomercury compounds and to remove the acetamidomethyl protecting group in organic synthesis. It is also involved in the oxymercuration reactions as well as to prepare dithiocarbonates from thiocarboantes.It finds application as a catalyst in organic synthesis, pharmaceuticals and absorption of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(II) acetate is used as a reagent to prepare organomercury compounds and to remove the acetamidomethyl protecting group in organic synthesis. It is also involved in the oxymercuration reactions as well as to prepare dithiocarbonates from thiocarboantes.It finds application as a catalyst in organic synthesis, pharmaceuticals and absorption of ethylene.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179°C to 182°C
Density 3.28
Linear Formula Hg(OOCCH3)2
Quantity 500 g
UN Number UN1629
Beilstein 3563831
Sensitivity Light sensitive
Merck Index 14,5873
Solubility Information Soluble in water.
Formula Weight 318.68
Percent Purity ≥98%
Chemical Name or Material Mercury(II) acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.