missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Acetate, ≥99.0% (Precipitation Titration), Solstice

Product Code. 15627270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15627270 50 g Poly bottle 50g
15617270 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15627270 Supplier Honeywell-Fluka Supplier No. 8335250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179°C to 182°C
Color Beige
Physical Form Powder
Quantity 50 g
Linear Formula (CH3COO)2Hg
UN Number UN1629
Beilstein 3563831
Formula Weight 318.68g/mol
Percent Purity ≥99.0% (precipitation titration)
Grade ACS/puriss. p.a.
Odor Characteristic
Packaging Poly bottle
Chemical Name or Material Mercury(II) acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.