missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Acetate, ACS Reagent, ≥98.0%, Honeywell Fluka™

Product Code. 15631570 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15631570 5 g Poly Bottle 5g
15611570 100 g Poly Bottle 100g
15621570 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15631570 Supplier Honeywell-Fluka Supplier No. 1761095G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number MFCD00012165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Melting Point 179°C to 182°C
Color White
Quantity 5 g
Linear Formula (CH3COO)2Hg
UN Number UN1629
Beilstein 3563831
Formula Weight 318.68g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 3.27g/cu.cm
Chemical Name or Material Mercury(Ii) Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.