missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Bromide, ≥99.0% (precipitation titration), Puriss. p.a., ACS Reagent, Honeywell™ Fluka™

Product Code. 15637270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15637270 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15637270 Supplier Honeywell-Fluka Supplier No. 8335350G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 322°C
Melting Point 237°C
CAS 7789-47-1
Color Violet to White
pH 4.5 (at 5.00g/L)
Physical Form Solid
Quantity 50 g
Molecular Formula Br2Hg
Linear Formula HgBr2
MDL Number MFCD00011040
UN Number UN1634
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
SMILES Br[Hg]Br
IUPAC Name dibromomercury
Molecular Weight (g/mol) 360.4
PubChem CID 24612
ChEBI CHEBI:49639
Formula Weight 360.4g/mol
Percent Purity ≥99.0% (precipitation titration)
Odor Odorless
Packaging Glass bottle
Vapor Pressure 1mmHg (136.5°C)
Density 6.1g/cu.cm at 25°C
Chemical Name or Material Mercury(II) bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Mercury dibromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.