missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Bromide, ACS Reagent, 98%, Honeywell™ Fluka™

Product Code. 15651720 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15651720 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15651720 Supplier Honeywell-Fluka Supplier No. 200085100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 322°C
Melting Point 237°C
CAS 7789-47-1
Color White to Yellow
pH 4.5 (at 5.00g/L)
Physical Form Solid
Quantity 100 g
Molecular Formula Br2Hg
Linear Formula HgBr2
MDL Number MFCD00011040
UN Number UN1634
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
SMILES Br[Hg]Br
IUPAC Name dibromomercury
Molecular Weight (g/mol) 360.4
PubChem CID 24612
ChEBI CHEBI:49639
Formula Weight 360.4g/mol
Percent Purity 98%
Grade ACS Reagent
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (136.5°C)
Density 6.1g/cu.cm at 25°C
Chemical Name or Material Mercury(Ii) Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.