missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) chloride, ACS, 99.5% min

Product Code. 11341619
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Artikelnummer. 11341619

missing translation for 'mfr': Thermo Scientific Alfa Aesar 012274.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol

Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Spezifikation

Boiling Point 302°C
Melting Point 280°C to 282°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% min.
Solubility Information Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides),alcohol,glycerol,methanol,acetone,and ethyl acetateSoluble in water,alcohol,acetone and ethyl acetate. Slightly soluble in benzene,carbon disulfide and pyridine.
Formula Weight 271.5
Grade ACS
Odor Odorless
Refractive Index 1.859
Appearance White powder
Vapor Pressure 0.0001 hPa
Density 5.4 g/cm3 at 20°C
Chemical Name or Material Mercury(II) chloride
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Mercury(II) chloride
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Gefahrenhinweis
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Mercury chloride

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt