missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Iodide Red, puriss., ≥99%, Honeywell Fluka™

Product Code. 15640940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Poly Bottle
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15640940 250 g Poly Bottle 250g
15630940 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15640940 Supplier Honeywell-Fluka Supplier No. 10015250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., red,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Boiling Point 354°C
Melting Point 259°C
Color Red
Quantity 250 g
UN Number UN1638
Formula Weight 454.4g/mol
Percent Purity ≥99%
Grade puriss.
Packaging Poly Bottle
Appearance Powder
Density 6.28g/cu.cm
Chemical Name or Material Mercury(Ii) Iodide Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.