missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Nitrate Monohydrate, ≥98%, Certified AR for Analysis, Fisher Chemical

Product Code. 11944281 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11944281

Brand: Fisher Chemical M/3012/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 77°C
CAS 7783-34-8
Color White
pH 3
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula H2HgN2O7
MDL Number MFCD00149736
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name mercury(2+) hydrate dinitrate
Molecular Weight (g/mol) 342.62
PubChem CID 3084029
Formula Weight 342.6g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) nitrate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.