missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Nitrate Monohydrate, purum p.a., ≥98.5% (T), Honeywell Fluka™

Product Code. 15617280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617280 10 g Poly Bottle 10g
15637280 50 g Poly Bottle 50g
15627280 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15617280 Supplier Honeywell-Fluka Supplier No. 8338110G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥98.5% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+);dinitrate;hydrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 79°C
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula Hg(NO3)2 · H2O
UN Number UN1625
Formula Weight 342.62g/mol
Percent Purity ≥98.5% (T)
Packaging Poly Bottle
Chemical Name or Material Mercury(II) nitrate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.