missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, 98+%

Product Code. 11427807
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427807 250 g 250g
11417807 50 g 50g
11407807 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427807 Supplier Thermo Scientific Alfa Aesar Supplier No. A16330.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-35-9 | HgO4S | 296.65 g/mol

It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Quantity 250 g
Linear Formula HgSO4
UN Number UN1645
Merck Index 14,5887
Solubility Information It is soluble in hot H2SO4,NaCl solution. insoluble in alcohol,acetone,ammonia
IUPAC Name mercury(2+) sulfate
Formula Weight 296.65
Percent Purity ≥98%
Sensitivity Moisture sensitive
Density 6.47
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.