missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Sulfate, Honeywell Fluka™

Product Code. p-7071753 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
Unit Size:
1kg
250g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15680940 250 g 250g
15670940 1 kg 1kg
15690940 6 x 250 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15680940 Supplier Honeywell-Fluka Supplier No. 10029250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Quantity 250 g
Linear Formula HgSO4
UN Number 1645
IUPAC Name mercury(2+);sulfate
Formula Weight 296.65g/mol
Percent Purity ≥98%
Grade puriss.
Chemical Name or Material Mercury(Ii) Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.