missing translation for 'onlineSavingsMsg'
Learn More

Meropenem Sodium Salt, TRC

Product Code. 17941915
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941915 5 mg 5mg
17974995 100 mg 100mg
17943695 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941915 Supplier Toronto Research Chemicals Supplier No. TRCM2256205MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211238-34-5
Molecular Formula C17 H24 N3 O5 S . Na
Molecular Weight (g/mol) 405.44
Synonym 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[[(3S,5S)-5-[(dimethylamino)carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-, sodium salt (1:1), (4R,5S,6S)- (ACI), 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[[(3S,5S)-5-[(dimethylamino)carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-, monosodium salt, (4R,5S,6S)- (9CI), (4R,5S,6S)-3-[[(3S,5S)-5-[(Dimethylamino)carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid sodium salt (1:1), Meropenem sodium, Meropenem sodium salt
IUPAC Name sodium;(4R,5S,6R)-3-[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
SMILES [Na+].C[C@@H](O)[C@H]1[C@H]2[C@@H](C)C(=C(N2C1=O)C(=O)[O-])S[C@@H]3CN[C@@H](C3)C(=O)N(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H25N3O5S.Na/c1-7-12-11(8(2)21)16(23)20(12)13(17(24)25)14(7)26-9-5-10(18-6-9)15(22)19(3)4;/h7-12,18,21H,5-6H2,1-4H3,(H,24,25);/q;+1/p-1/t7-,8-,9+,10+,11+,12-;/m1./s1
Formula Weight 405.1334
Percent Purity >95%
Chemical Name or Material Meropenem sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.