missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES hydrate, 99+%

Product Code. 15432958
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432958 250 g 250g
15442958 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432958 Supplier Thermo Scientific Chemicals Supplier No. H56472.30
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol

MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water. Slightly soluble in organic solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point ∽308°C (decomposition)
Quantity 250 g
Beilstein 141319
Merck Index 14,5902
Solubility Information Soluble in water. Slightly soluble in organic solvents.
Formula Weight 195.24 (Anhydrous)
Percent Purity ≥99%
Chemical Name or Material MES hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.