missing translation for 'onlineSavingsMsg'
Learn More

Mesembrine, TRC

Product Code. 17962565
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962565 5 mg 5mg
17963365 10 mg 10mg
17914155 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962565 Supplier Toronto Research Chemicals Supplier No. TRCM3389505MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24880-43-1
Molecular Formula C17H23NO3
Molecular Weight (g/mol) 289.37
Synonym (3aS,7aS)-3a-(3,4-Dimethoxyphenyl)octahydro-1-methyl-6H-indol-6-one, 4α,9α-Mesembrine, (3aS-cis)-3a-(3,4-Dimethoxyphenyl)octahydro-1-
SMILES COc1ccc(cc1OC)[C@]23CCN(C)[C@H]2CC(=O)CC3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H23NO3/c1-18-9-8-17(7-6-13(19)11-16(17)18)12-4-5-14(20-2)15(10-12)21-3/h4-5,10,16H,6-9,11H2,1-3H3
Formula Weight 289.1678
Percent Purity >95%
Chemical Name or Material Mesembrine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.