Learn More
3M™ Mesh Visor 5J-1
Product Code. 15769867 Shop All 3M Products
Change view
Click to view available options
Model: 5J-1
Product Code.
15769867 Model: 5J-1
1 options
Product Code. 15769867 Supplier 3M™ Supplier No. 7000103793
  • ONLINE EXCLUSIVE PRICE
    475.00 €
    339.37 € / Pack of 10
    Save 135.63 €
    28% Off
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
3M™ Mesh Visor 5J-1 is constructed of etched stainless steel mesh with a black matte finish and designed in 3 zones with different aperture sizes.
  • Etched Stainless Steel Visor
  • Optimal mesh design for enhanced stability and protection
  • Compatible with the 3M™ V5 Face Shield system for mounting on a 3M™ Hard Hats Series G3000 or G22
  • Snap in features makes it quick and easy to combine the face shield to the face shield holder
  • Optical class 1 visor, low energy impact, B
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN 1731
Thickness (Metric) 0.15 mm
Product Type Mesh Visor
For Use With (Application) Forestry
Model 5J-1
No. per Pack 10/Pk.
Material Etched Stainless Steel Mesh