missing translation for 'onlineSavingsMsg'
Learn More

Mesitoic acid, TRC

Product Code. 30135956
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135956 5 g 5g
17971705 1 g 1g
17940905 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135956 Supplier Toronto Research Chemicals Supplier No. TRCT8960405G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
Synonym 2,4,6-Trimethylbenzoic Acid, Mesitylenecarboxylic Acid, NSC 1119, NSC 203160;NSC 407983, beta-Isodurylic Acid
IUPAC Name 2,4,6-trimethylbenzoic acid
SMILES Cc1cc(C)c(C(=O)O)c(C)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12O2/c1-6-4-7(2)9(10(11)12)8(3)5-6/h4-5H,1-3H3,(H,11,12)
Formula Weight 164.0837
Chemical Name or Material Mesitoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.