missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene, TRC

Product Code. 30472029
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472029 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472029 Supplier Toronto Research Chemicals Supplier No. TRCM258390100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 1,3,5-Trimethylbenzene
CAS 108-67-8
Molecular Formula C9 H12
Quantity 100 mL
Synonym Benzene, 1,3,5-trimethyl-, Mesitylene (8CI), 1,3,5-Trimethylbenzene, 2,4,6-Trimethylbenzene, 3,5-Dimethyltoluene, NSC 9273, sym-Trimethylbenzene
InChI Formula InChI=1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3
SMILES Cc1cc(C)cc(C)c1
IUPAC Name 1,3,5-trimethylbenzene
Molecular Weight (g/mol) 120.19
Formula Weight 120.0939
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.