missing translation for 'onlineSavingsMsg'
Learn More

(Mesitylene)tungsten tricarbonyl, 98%, Thermo Scientific™

Product Code. 11324507
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11324507

Brand: Thermo Scientific Alfa Aesar 039589.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12129-69-0
Molecular Formula C12H12O3W
Molecular Weight (g/mol) 388.065
MDL Number MFCD00015319
InChI Key OENLQXWEAUYNBC-UHFFFAOYSA-N
Synonym tricarbonyl mesitylene tungsten 0, mesitylenetungsten tricarbonyl, tricarbonyl 1,2,3,4,5,6-eta-1,3,5-trimethylbenzene tungsten, tungsten,tricarbonyl 1,2,3,4,5,6-h-1,3,5-trimethylbenzene
PubChem CID 10981963
IUPAC Name carbon monoxide;1,3,5-trimethylbenzene;tungsten
SMILES CC1=CC(=CC(=C1)C)C.[C-]#[O+].[C-]#[O+].[C-]#[O+].[W]

Specifications

Melting Point 165°C (decomposition)
Quantity 250 mg
UN Number UN3466
Solubility Information Insoluble in water.
Formula Weight 388.08
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material (Mesitylene)tungsten tricarbonyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.