missing translation for 'onlineSavingsMsg'
Learn More

Mesna, TRC

Product Code. 30465137
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465137 Supplier Toronto Research Chemicals Supplier No. TRCM22575050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesna
CAS 19767-45-4
Molecular Formula C2 H5 O3 S2 . Na
Quantity 50 mg
Synonym Mesna, Sodium 2-sulfanylethanesulfonate, Ethanesulfonic acid, 2-mercapto-, sodium salt (1:1) (ACI), Ethanesulfonic acid, 2-mercapto-, monosodium salt (8CI, 9CI), 2-Mercapto-1-ethanesulfonic acid monosodium salt, 2-Mercaptoethanesulfonic acid monosodium salt, 2-Mercaptoethanesulfonic acid sodium salt, D 7093, MeSH ID: D015080, Mesnex, Mesnum, Mistabron, Mistabronco, Mitexan, Mucofluid, Prehepon, Sodium 2-mercaptoethanesulfonate, UCB 3983, Uromitexan
InChI Formula InChI=1S/C2H6O3S2.Na/c3-7(4,5)2-1-6;/h6H,1-2H2,(H,3,4,5);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)CCS
IUPAC Name sodium;2-sulfanylethanesulfonate
Molecular Weight (g/mol) 164.18
Formula Weight 163.9578
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.