missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific™

Product Code. 11455817
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
This item is not returnable. View return policy

Product Code. 11455817

Brand: Thermo Scientific Alfa Aesar B24382.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-36-6
Molecular Formula C4H2Br2O4
Molecular Weight (g/mol) 273.87
MDL Number MFCD00066439
InChI Key FJWGRXKOBIVTFA-XIXRPRMCSA-L
Synonym 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
PubChem CID 641611
IUPAC Name (2S,3R)-2,3-dibromobutanedioic acid
SMILES [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

Specifications

Melting Point ∼280°C
Quantity 100 g
UN Number UN3261
Beilstein 1725173
Merck Index 14,3029
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.
Formula Weight 275.89
Percent Purity 98%
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.