missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-Erythritol-1,1,2,3,4,4-d6, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Erythritol-d6 |
| CAS | 2714435-98-8 |
| Molecular Formula | C4 D6 H4 O4 |
| Quantity | 0.1 g |
| Synonym | (2 R,3 S)-rel-1,2,3,4-Butanetetrol-1,1,2,3,4,4-d6, rel-(2 R,3 S)-1,2,3,4-Butanetetrol-1,1,2,3,4,4-d6, Erythritol-1,1,2,3,4,4-D6, (2 R,3 S)-Butane-1,2,3,4-tetrol-1,2,3,4-13C4, meso-Erythritol-d6, meso-Erythritol-1,1,2,3,4,4-d6, Erythrit-d6, Erythritol-d6 |
| InChI Formula | InChI=1 S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8 H,1-2H2/t3-,4+/i1D2,2D2,3 D,4 D |
| SMILES | [2 H]C([2 H])(O)[C@]([2 H])(O)[C@]([2 H])(O)C([2 H])([2 H])O |
| IUPAC Name | (2 R,3 S)-1,1,2,3,4,4-hexadeuteriobutane-1,2,3,4-tetrol |
| Molecular Weight (g/mol) | 128.1568 |
| Formula Weight | 128.0956 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction