missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol, 99%

Product Code. 11402488
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402488 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402488 Supplier Thermo Scientific Alfa Aesar Supplier No. A15813.36
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol

meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

Solubility
Soluble in water (0.1 g/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
TRUSTED_SUSTAINABILITY

Specifications

CAS 149-32-6
Melting Point 119°C to 124°C
Boiling Point 329°C to 331°C
Assay Percent Range 99%
Molecular Formula C4H10O4
MDL Number MFCD00004710
Quantity 500 g
Beilstein 1719753
Merck Index 14,3675
Synonym erythritol, meso-erythritol, phycitol, erythrit, phycite
Solubility Information Soluble aq. soln. (0.1 g/ml).
InChI Key UNXHWFMMPAWVPI-UHFFFAOYNA-N
SMILES OCC(O)C(O)CO
IUPAC Name butane-1,2,3,4-tetrol
Molecular Weight (g/mol) 122.12
PubChem CID 222285
ChEBI CHEBI:17113
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material meso-Erythritol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.