missing translation for 'onlineSavingsMsg'
Learn More

Mestranol, TRC

Product Code. 30482790
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482790 500 mg 500mg
30479193 10 mg 10mg
30480448 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482790 Supplier Toronto Research Chemicals Supplier No. TRCM258755500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-33-3
Molecular Formula C21 H26 O2
Molecular Weight (g/mol) 310.43
Synonym Mestranol, 3-methoxy-19-nor-17α-pregna-1,3,5(10)-trien-20-yn-17-ol, 19-Nor-17α-pregna-1,3,5(10)-trien-20-yn-17-ol, 3-methoxy- (6CI,8CI), 17-Ethynyl-3-methoxy-1,3,5(10)-estratrien-17β-o1, 17-Ethynyl-3-methoxyestra-1,3,5(10)-trien-17β-ol, 17-Ethynylestradiol 3-methyl ether, 17α-Ethinylestradiol 3-methyl ether, 17α-Ethynyl-3-methoxy-1,3,5(10)-estratrien-17β-ol, 17α-Ethynyl-3-methoxy-17β-hydroxy-δ1,3,5(10)-estratriene, 17α-Ethynylestradiol 3-methyl ether, 17α-Ethynylestradiol methyl ether, 3-Methoxy-17α-ethinylestradiol, 3-Methoxy-17α-ethynylestradiol, 3-Methoxy-19-nor-17α-pregna-1,3,5(10)-trien-20-yn-17-ol, 3-Methoxy-19-nor-17α-pregna-1,3,5(10)-trien-20-yn-17β-ol, 3-Methoxyethynylestradiol, 3-Methylethynylestradiol, 3-O-Methyl-17α-ethynylestradiol, 3-O-Methylethynylestradiol, 33355, 8027 C. B., Devocin, EE 3ME, Ethinylestradiol 3-methyl ether, Ethynylestradiol 3-methyl ether, Ethynylestradiol 3-monomethyl ether, Ethynylestradiol methyl ether, Inostral, Menophase, Mestranol, NSC 84032, Norquen, Ovastol, SC 4725, δ-MVE, 19-Norpregna-1,3,5(10)-trien-20-yn-17-ol, 3-methoxy-, (17α)-
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-3-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-ol
SMILES COc1ccc2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCc2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H26O2/c1-4-21(22)12-10-19-18-7-5-14-13-15(23-3)6-8-16(14)17(18)9-11-20(19,21)2/h1,6,8,13,17-19,22H,5,7,9-12H2,2-3H3/t17-,18-,19+,20+,21+/m1/s1
Formula Weight 310.1933
Chemical Name or Material Mestranol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.