missing translation for 'onlineSavingsMsg'
Learn More

Met-enkephalin, TRC

Product Code. 30007843
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007843 25 mg 25mg
30004116 100 mg 100mg
30007263 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007843 Supplier Toronto Research Chemicals Supplier No. TRCM22548525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58569-55-4
Synonym Human β-Endorphin(1-5)∣L-Methionine-enkephalin∣Lupex∣Met-enkephalin∣Met5-enkephalin∣Metenkefalin∣Methionine Enkephalin∣Methionyl-enkephalin∣NSC 374896∣Opioid Growth Factor∣PN: US5961923 PAGE: 135 claimed protein∣Peptid-M∣53: PN: US20090104234 SEQID: 53 unclaimed protein∣5: PN: US20080039404 SEQID: 5 unclaimed sequence∣6: PN: US20130072438 SEQID: 5 unclaimed sequence∣6: PN: WO0130371 TABLE: 1 unclaimed sequence∣707: PN: US20090175821 SEQID: 882 claimed protein∣7: PN: US20130303729 SEQID: 7 unclaimed protein∣7: PN: WO0130371 TABLE: 1 unclaimed sequence∣7: PN: WO2008106043 TABLE: 6 claimed protein∣9: PN: WO03061683 FIGURE: 1 unclaimed sequence∣9: PN: WO2004041151 SEQID: 9 unclaimed sequence∣5-L-Methionine-enkephalin∣5-Methionine Enkephalin∣105: PN: US20030119021 SEQID

Specifications

Format Neat
Molecular Weight (g/mol) 573.66
Product Type API
Quantity 25 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.