missing translation for 'onlineSavingsMsg'
Learn More

meta-Chlorambucil, TRC

Product Code. 30362136
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362136 5 mg 5mg
30362087 25 mg 25mg
30357785 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362136 Supplier Toronto Research Chemicals Supplier No. TRCC3251305MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134862-11-6
Molecular Formula C14H19Cl2NO2
Molecular Weight (g/mol) 304.21
Synonym 3-[Bis(2-chloroethyl)amino]benzenebutanoic Acid, Chlorambucil Impurity
IUPAC Name 4-[3-[bis(2-chloroethyl)amino]phenyl]butanoic acid
SMILES OC(=O)CCCc1cccc(c1)N(CCCl)CCCl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H19Cl2NO2/c15-7-9-17(10-8-16)13-5-1-3-12(11-13)4-2-6-14(18)19/h1,3,5,11H,2,4,6-10H2,(H,18,19)
Chemical Name or Material meta-Chlorambucil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.