missing translation for 'onlineSavingsMsg'
Learn More

Edulab Metal Density Cubes - Set of 7

Product Code. 30280279
Change view
Click to view available options
Product Type:
Density Cubes
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30280279 Density Cubes 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280279 Supplier Edulab Supplier No. VTN12302208
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A complete set of seven labelled metal cubes, ideal for classroom demonstrations and density investigations.

  • Includes seven labelled metal cubes made of brass, lead, zinc, copper, aluminum, iron, and tin
  • Designed for hands-on classroom experiments exploring density and material properties
  • Each cube provides consistent results under standard conditions of temperature and pressure
  • Helps students understand how density relates to mass and volume in practical applications
  • Supplied in a durable plastic storage case for organization and protection
  • Well-suited for physics lessons to compare predictable densities across different metals

Specifications

Grade 9+
Format Set
Product Type Density Cubes
Languages English
Subject Physics

Prop 65 compliance: On Product Combined Cancer Reproductive

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.