Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-413
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
Brand:  Brady™ 104169
 View more versions of this product
Product Code. 11579413
  • ONLINE EXCLUSIVE PRICE
    465.00 €
    448.32 € / 1 rolls
    Save 16.68 €
    3% Off
In Stock
Add to basket
Description
Description
  • Available in various sizes
  • Acrylic-based, industrial-strength adhesive for multiple applications
Specifications
Specifications
UL Recognised; cUL Recognised
Acrylic, Permanent
Light Gray
Roll
Single-side Printable
Label
Asset Tracking, General and Industrial Identificationentification, Rating Plates
27.31 mm
-70°C
10000 Label(s) / Roll
Rectangle
25.4 x 9.52 mm (W x H)
-70°C to +110°C
0.086 mm
9.53 x 25.40 mm
1 in.
25.40 mm
9.52 mm
Thermal Transfer Printable Labels
None
Satin
Polyester (B-480)
Thermal Transfer
Non-sterile
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
3
THT-3-413-10
10,000/Pk.
25.40 mm (W) x 9.53 mm (H)
-94°F to +230°F
0.0034 in.
12.07 mm
0.375 in.
25.4 mm
0.375 in.
THT-3-413-10