Learn More
Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes
Product Code. 12966986 Shop All Nickel-Electro Products
Click to view available options
Length (Metric):
300 mm
340 mm
400 mm
430 mm
490 mm
Unit Size:
Each
Product Code. 12966986
Brand: Nickel Electro™ 6702/SS
Only null left
Add to basket
Add to basket
Keep pipets sterile with a sterilizing box. Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes can be steam sterilized at 120°C and withstand dry heat temperatures of up to 180°C.
  • Stainless steel square profile
  • Includes lid
  • Square shape prevents it from rolling off bench top
  • Silicon in lid and base
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Sterility Sterile
Quantity Each
Length (Metric) 340 mm
Type Sterilizing Box
For Use With (Equipment) Autoclave