missing translation for 'onlineSavingsMsg'
Learn More

Metamizole Magnesium Salt, TRC

Product Code. 30006317
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006317 2.5 g 2.5g
30006172 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006317 Supplier Toronto Research Chemicals Supplier No. TRCM2256602.5G
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63372-86-1
Molecular Formula 2 C13 H16 N3 O4 S . Mg
Molecular Weight (g/mol) 645
Synonym Magnesium, bis[[(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino-κN]methanesulfonato-κO]-, (T-4)- (9CI), Magnesium, bis[[(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]methanesulfonato]-, (T-4)-, Methanesulfonic acid, [(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-, magnesium complex, Magnalgin, Magnesium dipyrone, Metamizole Magnesium complex
IUPAC Name magnesium;[(1,5-dimethyl-3-oxo-2-phenyl-pyrazol-4-yl)methylamino]methanesulfonate
SMILES [Mg+2].CN1N(C(=O)C(=C1C)CNCS(=O)(=O)[O-])c2ccccc2.CN3N(C(=O)C(=C3C)CNCS(=O)(=O)[O-])c4ccccc4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/2C13H17N3O4S.Mg/c2*1-10-12(8-14-9-21(18,19)20)13(17)16(15(10)2)11-6-4-3-5-7-11;/h2*3-7,14H,8-9H2,1-2H3,(H,18,19,20);/q;;+2/p-2
Formula Weight 644.1573
Chemical Name or Material Metamizole Magnesium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.