missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metanil Yellow

Product Code. 11488147
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11488147 250 g 250g
11478147 50 g 50g
2 options
This item is not returnable. View return policy

Product Code. 11488147

Brand: Thermo Scientific Chemicals A17527.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol

Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

Solubility
Soluble in water (25g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.378
MDL Number MFCD00007486
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Synonym C.I. 13065
PubChem CID 3935589
ChEBI CHEBI:87235
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

Beilstein 3831568
Merck Index 14,5928
Solubility Information Soluble in water (25g/L).
Formula Weight 375.38
Odor Odorless
pH 10
Quantity 250 g
Chemical Name or Material Metanil Yellow
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.