missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metanil Yellow

Produktkod. 10730872 Handla allt< titel> Produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
Förpackningsstorlek
1kg
Produktkod. Quantity unitSize
10730872 1 kg 1kg
1 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 10730872

missing translation for 'mfr': Thermo Scientific Chemicals 413720010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 587-98-4
Dye Content 88% min.
Molecular Formula C18H14N3NaO3S
Synonym Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in benzene and ether, slightly soluble in acetone
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Infrared Spectrum Authentic
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
Molecular Weight (g/mol) 375.378
PubChem CID 3935589
ChEBI CHEBI:87235
Lambda Max 413 to 419nm (in methanol)
Formula Weight 375.38
Packaging Glass bottle
Quantity 1 kg
Chemical Name or Material Metanil Yellow
Visa mer Visa mindre
compliance-icons compliance-icons
Signalord
  • Warning
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Faroangivelser
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.