missing translation for 'onlineSavingsMsg'
Learn More

Metanil Yellow, 98%, pure, ACROS Organics™

Product Code. 13438438 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
13438438 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 13438438

Brand: Thermo Scientific Acros 191965000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Indicator
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 550 (at lambda min.)
CAS 587-98-4
Absorption Ratio 0.9 to 1.1
Assay Percent Range 98%
Molecular Formula C18H14N3NaO3S
MDL Number MFCD00007486
Synonym Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Infrared Spectrum Authentic
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
Molecular Weight (g/mol) 375.378
PubChem CID 3935589
ChEBI CHEBI:87235
Lambda Max 431 to 436nm (in water)
Formula Weight 375.38
Loss on Drying 10% max. (105°C)
Percent Purity ≥97.5%
Grade Pure
Packaging Glass bottle
Color Orange
Quantity 500 g
Physical Form Powder
Chemical Name or Material Metanil Yellow, 98%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.