missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric Acid Potassium Salt, TRC

Product Code. 30477590
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477590 25 g 25g
30467571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477590 Supplier Toronto Research Chemicals Supplier No. TRCM22626025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metaphosphoric Acid Potassium Salt
CAS 7790-53-6
Molecular Formula HKO3P
Quantity 25 g
Synonym Potassium Metaphosphate, Kurrol's Salt, Potassium Kurrol's Salt, Potassium Metaphosphate, Potassium Phosphate (KPO3), Potassium Polymetaphosphate, Phosphenic Acid Potassium Salt
InChI Formula InChI=1S/K.HO3P/c;1-4(2)3/h;(H,1,2,3)/q+1;/p-1
SMILES O=P(O[K])=O
Molecular Weight (g/mol) 118.07
Formula Weight 117.9222
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.