missing translation for 'onlineSavingsMsg'
Learn More

Metergoline, TRC

Product Code. 17951635
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951635 25 mg 25mg
17973345 250 mg 250mg
17941435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951635 Supplier Toronto Research Chemicals Supplier No. TRCM22627025MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17692-51-2
Molecular Formula C25 H29 N3 O2
Molecular Weight (g/mol) 403.5167
Synonym N-[[(8β)-1,6-Dimethylergolin-8-yl]methyl]-carbamic Acid Phenylmethyl Ester, Carbamic acid, [(1,6-dimethylergolin-8β-yl)methyl]-, benzyl ester (8CI), Carbamic acid, [[(8β)-1,6-dimethylergolin-8-yl]methyl]-, phenylmethyl ester (9CI), Lysergamine, N-carboxy-9,10-dihydro-1-methyl-, benzyl ester (7CI), Ergoline, carbamic acid deriv., Indolo[4,3-fg]quinoline, carbamic acid deriv., (+)-N-Carboxy-1-methyl-9,10-dihydrolysergamide benzyl ester, 8β-Carbobenzyloxyaminomethyl-1,6-dimethyl-10α-ergoline, AHR 3009, Contralac, D-8β-[(Carbobenzoxyamino)methyl]-1,6-dimethyl-10α-ergoline, D-8β-[(Carboxyamino)methyl]-1,6-dimethylergoline I benzyl ester, D-N-Carbobenzoxydihydro-1-methyllysergamine I, D-N-Carboxydihydro-1-methyllysergamine I benzyl ester, D-[(4,6,6a,7,8,9,10,10a-Octahydro-4,7-dimethyl-10aα-indolo[4,3-fg]quinolin-9β-yl)methyl]carbamic acid benzyl ester, FI 6337, Liserdol, MCE, Metergolin, Methergoline, [(1,6-Dimethylergolin-8β-yl)methyl]carbamic Acid Benzyl Ester, N-Carboxy-9,10-dihydro-1-methyllysergamine Benzyl Ester
IUPAC Name benzyl N-[[(6aR,9S,10aR)-4,7-dimethyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-9-yl]methyl]carbamate
SMILES CN1C[C@H](CNC(=O)OCc2ccccc2)C[C@H]3[C@H]1Cc4cn(C)c5cccc3c45

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H29N3O2/c1-27-14-18(13-26-25(29)30-16-17-7-4-3-5-8-17)11-21-20-9-6-10-22-24(20)19(12-23(21)27)15-28(22)2/h3-10,15,18,21,23H,11-14,16H2,1-2H3,(H,26,29)/t18-,21+,23+/m0/s1
Formula Weight 403.226
Chemical Name or Material Metergoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.