missing translation for 'onlineSavingsMsg'
Learn More

MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™

Product Code. 11472432 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472432 5 g 5g
11462432 1 g 1g
11482432 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472432 Supplier MP Biomedicals Supplier No. 0215169105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
PubChem CID 14219
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

Color White
Quantity 5 g
Formula Weight 165.6
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Metformin hydrochloride, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.