missing translation for 'onlineSavingsMsg'
Learn More

Metformin Hydrochloride, TRC

Product Code. 17995085
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17995085 1 g 1g
17992235 5 g 5g
17953095 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17995085 Supplier Toronto Research Chemicals Supplier No. TRCM2588151G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4 H11 N5 . Cl H
Molecular Weight (g/mol) 165.62
Synonym Imidodicarbonimidic diamide, N,N-dimethyl-, hydrochloride (1:1), Biguanide, 1,1-dimethyl-, hydrochloride (6CI), Biguanide, 1,1-dimethyl-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N,N-dimethyl-, monohydrochloride (9CI), 1,1-Dimethylbiguanide hydrochloride, Apo-Metformin, Apophage, Benofomin, Brot, Dabex, Denkaform, Dextin, Diabefagos, Diabetmim, Diabetmin, Diabetosan, Diabex, Diaformin, Dialon, Diformin, Diformin Retard, Dimefor, FORTAMET, Fornidd, Geamet, Glifage, Glucaminol, Glucofago, Glucoform, Glucoformin, Glucomet, Glucomin, Glucomine, Gluconil, Glucophage, Glucophage 850, Glucophage Forte, Glucophage Retard, Glucophage SR, Glucophage XR, Glucophage-Mite, Gludepatic, Glufor, Gluformin, Glumeformin, Glumetza, Glumin, Glumin XR, Glupermin, Glyceriphage, Glyciphage, Glycon, Glyformin, LA 6023, Meguan, Melbin, Metforal, Metformin hydrochloride, Metgluco, Metomin, Miformin, N,N-Dimethylbiguanide hydrochloride, N,N-Dimethylbiguanidine monohydrochloride, N,N-Dimethylbiguanidinium chloride, N1,N1-Dimethylbiguanide hydrochloride, Neodipa, Orabet, Riomet, Siamformet, Walaphage
IUPAC Name 3-carbamimidoyl-1,1-dimethylguanidine;hydrochloride
SMILES Cl.CN(C)C(=N)NC(=N)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H
Formula Weight 165.0781
Percent Purity >95%
Chemical Name or Material Metformin Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.