missing translation for 'onlineSavingsMsg'
Learn More

Metformin, TRC

Product Code. 30010834
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010834 100 mg 100mg
30006001 10 mg 10mg
30005677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010834 Supplier Toronto Research Chemicals Supplier No. TRCM258858100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 657-24-9
Molecular Formula C4 H11 N5
Molecular Weight (g/mol) 129.16
Synonym Imidodicarbonimidic diamide, N,N-dimethyl-, Biguanide, 1,1-dimethyl- (6CI,8CI), N,N-Dimethylimidodicarbonimidic diamide, 1,1-Dimethylbiguanide, Cidophage, DMGG, Dimethylbiguanide, Emiphage, Ficonax, Fluamine, Flumamine, Glifor, Gliguanid, Glucofine, Haurymelin, Metformin, Metiguanide, Metphage, N,N-Dimethylbiguanide, N,N-Dimethyldiguanide, N1,N1-Dimethylbiguanide, NNDG, N'-Dimethylguanylguanidine, Siofor
IUPAC Name 3-carbamimidoyl-1,1-dimethylguanidine
SMILES CN(C)C(=N)NC(=N)N

Especificaciones

Quantity 100 mg
InChI Formula InChI=1S/C4H11N5/c1-9(2)4(7)8-3(5)6/h1-2H3,(H5,5,6,7,8)
Formula Weight 129.1014
Percent Purity >95%
Chemical Name or Material Metformin
Grade HPLC
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.