missing translation for 'onlineSavingsMsg'
Learn More

Methacholine chloride, 100.4%, MP Biomedicals™

Product Code. 11421691 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
This item is not returnable. View return policy

Product Code. 11421691

Brand: MP Biomedicals 0219023105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62-51-1
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.687
MDL Number MFCD00011817
InChI Key JHPHVAVFUYTVCL-UHFFFAOYSA-M
Synonym methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride
PubChem CID 6114
ChEBI CHEBI:50142
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)C.[Cl-]

Specifications

Color White
Assay Percent Range ≥98%
Quantity 5 g
Formula Weight 195.7
Physical Form Powder
Chemical Name or Material Methacholine chloride
compliance-icons
Product Identifier
  • Methacholine Chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methacholine chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.