missing translation for 'onlineSavingsMsg'
Learn More

Methacrolein, 90%, stabilized, AcroSeal™

Product Code. 10063907 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10063907 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10063907 Supplier Thermo Scientific Acros Supplier No. 196501000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-85-3 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-85-3
Melting Point -81.0°C
Color Colorless to Yellow
Density 0.8470g/mL
Boiling Point 69.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H6O
Linear Formula H2C=C(CH3)CHO
Refractive Index 1.4145
MDL Number MFCD00006974
Quantity 100 mL
Beilstein 01,III,2981
Specific Gravity 0.847
Synonym methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
Solubility Information Solubility in water: 6g/100mL (20°C).
InChI Key STNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILES CC(=C)C=O
IUPAC Name 2-methylprop-2-enal
Stabilizer 0.1% hydroquinone
Molecular Weight (g/mol) 70.09
PubChem CID 6562
Viscosity 0.49 mPa.s (20°C)
Formula Weight 70.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methacrolein, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.