missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methacrylamide, 98%, extra pure

Product Code. 10366500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10366500 250 g 250g
10285240 500 g 500g
10703075 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366500 Supplier Thermo Scientific Chemicals Supplier No. 149202500
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 79-39-0 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Specifications

Melting Point 108.0°C to 112.0°C
Color White to Yellow
Boiling Point 215.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CONH2
Quantity 250 g
Beilstein 02, II, 399
Solubility Information Solubility in water: 202g/L (20°C)
Sublimation Point 0095.0°C
Formula Weight 85.11
Percent Purity 98%
Grade Extra Pure
Physical Form Crystals
Chemical Name or Material Methacrylamide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methacrylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.