missing translation for 'onlineSavingsMsg'
Learn More

Methacrylamide, TRC

Product Code. 30474045
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474045 2.5 g 2.50g
30471348 250 mg 250mg
30465810 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474045 Supplier Toronto Research Chemicals Supplier No. TRCM2588132.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4 H7 N O
Molecular Weight (g/mol) 85.1
Synonym 2-Propenamide, 2-methyl- (9CI, ACI), 2-Methyl-2-propenamide (ACI), Methacrylamide (8CI), 2-Methylacrylamide, 2-Methylpropenamide, Methacrylic acid amide, Methacrylic amide, NSC 23772, NSC 24147
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C4H7NO/c1-3(2)4(5)6/h1H2,2H3,(H2,5,6)
Formula Weight 85.0528
Chemical Name or Material Methacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.