missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic acid, 99%, stab. with 100-250ppm hydroquinone or 4-methoxyphenol

Product Code. 11464563
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
Product Code. Quantity unitSize
11464563 500 g 500g
11474563 2500 g 2500g
2 options
This item is not returnable. View return policy

Product Code. 11464563

Brand: Thermo Scientific Alfa Aesar A11543.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-41-4 | C4H6O2 | 86.09 g/mol

Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

Solubility
Miscible with warm water, chloroform, ethanol, ether and organic solvents.

Notes
Incompatible with amines, strong bases, strong acids, oxidizing agents and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-41-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002651
InChI Key CERQOIWHTDAKMF-UHFFFAOYSA-N
Synonym methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
PubChem CID 4093
ChEBI CHEBI:25219
IUPAC Name 2-methylprop-2-enoic acid

Specifications

Melting Point 16°C
Density 1.015
pH 2.0 to 2.2
Boiling Point 163°C
Flash Point 65°C (149°F)
Odor Acrid
Refractive Index 1.431
Quantity 500 g
UN Number UN2531
Beilstein 1719937
Sensitivity Light and moisture sensitive
Merck Index 14,5941
Solubility Information Miscible with warm water,chloroform,ethanol,ether and organic solvents.
SMILES CC(=C)C(=O)O
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material Methacrylic acid, Stabilized with 100 to 250ppm hydroquinone or 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methacrylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methacrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.