missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol

Product Code. 11377128
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377128 3 kg 3kg
11357128 250 g 250g
11367128 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377128 Supplier Thermo Scientific Alfa Aesar Supplier No. 015546.A5
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 79-41-4 | C4H6O2 | 86.09 g/mol

Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly (methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-41-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002651
InChI Key CERQOIWHTDAKMF-UHFFFAOYSA-N
Synonym methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
PubChem CID 4093
ChEBI CHEBI:25219
IUPAC Name 2-methylprop-2-enoic acid
SMILES CC(=C)C(=O)O

Specifications

Melting Point 16°C
Density 1.015 g/mL
pH 2.0 to 2.2
Boiling Point 163°C
Flash Point 65°C (149°F)
Assay Percent Range >99%
Odor Acrid
Refractive Index 1.431
Quantity 3 kg
UN Number UN2531
Beilstein 1719937
Sensitivity Light and moisture sensitive
Merck Index 14,5941
Solubility Information Miscible with warm water,chloroform,ethanol,ether and organic solvents.
Formula Weight 86.09
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Methacrylic acid, Stabilized with 250ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.