missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic Acid And Methyl Methacrylate Copolymer, TRC

Product Code. 30476215
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476215 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476215 Supplier Toronto Research Chemicals Supplier No. TRCM22594850G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methacrylic Acid Copolymer Type C
CAS 25086-15-1
Molecular Formula ((C5 H8 O2)mon(C4 H6 O2)mon)co
Quantity 50 g
Synonym Methacrylic Acid Copolymer Type A, Methacrylic Acid Copolymer Type B, Methacrylic Acid Copolymer Type C, 2-Propenoic acid, 2-methyl-, polymer with methyl 2-methyl-2-propenoate, Methacrylic acid, polymer with methyl methacrylate (8CI), 2-Propenoic acid, 2-methyl-, methyl ester, polymer with 2-methyl-2-propenoic acid (9CI), Methacrylic acid methyl ester, polymer with methacrylic acid (8CI), AR/P 610, AST 1, Acronal A 508, Acrypet MF 101, Altuglas HT 121, CP 19Y, Carboset XPD 1034, Carboset XPD 1234, Carboset XPD 1708, Carboset XPD 1708G, Copolymer of methylmethacrylate with methacrylic acid, Drugcoat L 100, Drugcoat S 100, Elvacite 1040, Elvacite 4055, Eudispert, Eudispert HV, Eudispert MV, Eudispert NV, Eudragit L, Eudragit L 100, Eudragit L 12.5, Eudragit S 100, Eudragit S 12.5, F 3031, F 303L, HL 370, KL 7177, M 14, M 14 (acrylic polymer), MMA8.5MAA-EL11, Metakril 14, Methacrylate copolymer L, Methacrylic acid-methyl methacrylate copolymer, Methacrylic acid-methyl methacrylate polymer, Methacrylic-methyl methacrylate polymers, Methyl methacrylate copolymer with methacrylic acid, Methyl methacrylate-methacrylic acid copolymer, Methyl methacrylate-methacrylic acid copolymers, Methyl methacrylate-methacrylic acid polymer, Methyl methacrylate-methacrylic acid polymers, OV 5300, P 118, P 118 (acrylic polymer), Poly(methacrylic acid-co-methyl methacrylate), Poly(methacrylic acid-methyl methacrylate), Poly(methyl methacrylate-co-methacrylic acid), Poly(methyl methacrylate-methacrylic acid), Rohagit S, Rohagit S-ENV, Rohagit SLV, S 100, SBP 5, SBP 5 (acrylic polymer), SOSK 102, SOSK 113.5, SOSZ 0.33.5, Synocryl 841S, Vitam 2M, Vitan 2M, XPD 1234
SMILES CC(=C)C(=O)O.CC(=C)C(=O)OC
Molecular Weight (g/mol) 186.21
Formula Weight 186.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.