missing translation for 'onlineSavingsMsg'
Learn More

Methacryloyl chloride, 95%, contains 200 ppm MEHQ as stabilizer, AcroSeal™, Thermo Scientific™

Product Code. 10655222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
AcroSeal™ Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10655222 1 L AcroSeal™ Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10655222 Supplier Thermo Scientific Acros Supplier No. 168660010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-46-7
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl

Specifications

Melting Point -60.0°C
Density 1.0760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H5ClO
Linear Formula H2C=C(CH3)COCl
Refractive Index 1.4447
Quantity 1 L
Beilstein 02,II,394
Specific Gravity 1.076
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common inert organic solvents,like ether,acetone and chloroform
Stabilizer 200ppm MEHQ max.
Molecular Weight (g/mol) 104.54
Formula Weight 104.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methacryloyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methacryloyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.