missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacycline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Methacycline Hydrochloride |
| CAS | 3963-95-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H22 N2 O8 . Cl H |
| Quantity | 25 g |
| Synonym | (4S,4aR,5S,5aR,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, (4S,4aR,5S,5aR,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5α,5aα,12aα)]-, 6-Demethyl-6-deoxy-5-hydroxy-6-methylenetetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methylene-5-hydroxytetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride, 6-Methylene-5-hydroxytetracycline chlorohydrate, 6-Methylene-5-hydroxytetracycline hydrochloride, 6-Methyleneoxytetracycline hydrochloride, Adriamicina, Ciclobiotic, Germiciclin, Londomycin, Metacyclin hydrochloride, Metacycline hydrochloride, Metadomus, Methacycline hydrochloride, Methacycline monohydrochloride, Metilenbiotic, Optimycin, Physiomycine, Rindex, Doxycycline Monohydrate Imp. B (EP) as Hydrochloride |
| InChI Formula | InChI=1S/C22H22N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-6,10,14-15,17,25,27-29,32H,1H2,2-3H3,(H2,23,31);1H/t10-,14-,15+,17+,22+;/m1./s1 |
| SMILES | Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@@H]3C(=C)c4cccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N |
| IUPAC Name | (4S,4aR,5S,5aR,12aS)-4-(dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methylidene-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 478.88 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?