missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonamide, 97+%

Product Code. 11424243
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
Unit Size:
10g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424243 500 g 500g
11478820 10 g 10g
11488820 50 g 50g
11498820 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11424243 Supplier Thermo Scientific Alfa Aesar Supplier No. A10885.36
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3144-09-0 | CH5NO2S | 95.12 g/mol

Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.

Solubility
Slightly soluble in DMSO and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3144-09-0
Molecular Formula CH5NO2S
Molecular Weight (g/mol) 95.12
MDL Number MFCD00007940
InChI Key HNQIVZYLYMDVSB-UHFFFAOYSA-N
Synonym methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide
PubChem CID 72879
IUPAC Name methanesulfonamide
SMILES CS(N)(=O)=O

Specifications

Melting Point 87°C to 93°C
Quantity 500 g
Beilstein 1740835
Solubility Information Slightly soluble in DMSO and methanol.
Formula Weight 95.13
Percent Purity ≥98%
Chemical Name or Material Methanesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.